Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

361571

Sigma-Aldrich

CHIR99021

≥95% (HPLC), liquid, GSK-3 inhibitor, Calbiochem

Synonym(s):

InSolution GSK-3 Inhibitor XVI, CHIR99021, 6-(2-(4-(2,4-Dichlorophenyl)-5-(4-methyl-1H-imidazol-2-yl)-pyrimidin-2-ylamino)ethyl-amino)-nicotinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18Cl2N8
CAS Number:
Molecular Weight:
465.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

product name

GSK-3 Inhibitor XVI, CHIR99021, InSolution, ≥95%, 25 mM

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−70°C

InChI

1S/C22H18Cl2N8/c1-13-10-29-21(31-13)17-12-30-22(32-20(17)16-4-3-15(23)8-18(16)24)27-7-6-26-19-5-2-14(9-25)11-28-19/h2-5,8,10-12H,6-7H2,1H3,(H,26,28)(H,29,31)(H,27,30,32)

InChI key

AQGNHMOJWBZFQQ-UHFFFAOYSA-N

General description

A cell-permeable aminopyrimidine compound that acts as potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC50 = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively). Exhibits little or no activity toward 46 other enzymes (IC50 >8.8 µM against 20 kinases and Ki ≥5 µM against 23 non-kinase enzymes) and shows only weak binding to 22 pharmacologically relevant receptors (Kd ~4 µM). This compound is orally available and is shown to effectively improve glucose metabolism in both murine and rat type 2 diabetes models in vivo. The solid form of this compound (Cat. No. 361559) is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GSK-3 α and β
Reversible: yes
Target IC50: 10 and 6.7 nM against GSK-3&alpha

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 25 mM (5 mg/430 µL) solution of GSK-3 Inhibitor XVI, CHIR99021 (Cat. No. 361559) in DMSO.

Other Notes

Mussmann, R., et al. 2007. J. Biol. Chem.282, 12030.
Ring, D.B., et al. 2003. Diabetes52, 588.
Bennett, C.N., et al. 2002. J. Biol. Chem.277, 30998.
Cline, G.W., et al. 2002.
Diabetes51, 2903.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanxin Fan et al.
STAR protocols, 4(1), 102073-102073 (2023-03-01)
Mitochondrial metabolism is critical in hematopoietic stem cell maintenance and differentiation. Here, we present a step-by-step protocol to efficiently differentiate human induced pluripotent stem cells into myeloid progenitors by a robust feeder- and serum-free system. Furthermore, we provide a protocol
Sunanjay Bajaj et al.
The EMBO journal, 40(23), e108714-e108714 (2021-10-19)
Inhibitory GABAergic interneurons migrate over long distances from their extracortical origin into the developing cortex. In humans, this process is uniquely slow and prolonged, and it is unclear whether guidance cues unique to humans govern the various phases of this
Gayan I Balasooriya et al.
Nature communications, 13(1), 5984-5984 (2022-10-11)
Each mammalian autosomal gene is represented by two alleles in diploid cells. To our knowledge, no insights have been made in regard to allele-specific regulatory mechanisms of autosomes. Here we use allele-specific single cell transcriptomic analysis to elucidate the establishment
Daniel Reumann et al.
Nature methods, 20(12), 2034-2047 (2023-12-06)
Ventral midbrain dopaminergic neurons project to the striatum as well as the cortex and are involved in movement control and reward-related cognition. In Parkinson's disease, nigrostriatal midbrain dopaminergic neurons degenerate and cause typical Parkinson's disease motor-related impairments, while the dysfunction
Yanfei Deng et al.
Cellular reprogramming, 22(4), 217-225 (2020-07-17)
Wnt/β-Catenin signaling pathway plays an important role in maintaining self-renewal and pluripotency of human and mouse embryonic stem cells (ESCs). Activation of Wnt/β-Catenin signaling pathway by glycogen synthase kinase-3 (GSK3) inhibitor, the Wnt signaling agonist, could maintain the pluripotency of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service