Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1285931

USP

Fluvastatin sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25FNNaO4
CAS Number:
Molecular Weight:
433.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluvastatin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].CC(C)n1c(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)c(-c2ccc(F)cc2)c3ccccc13

InChI

1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m0./s1

InChI key

ZGGHKIMDNBDHJB-RPQBTBOMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluvastatin sodium USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Fluvastatin Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Sakai et al.
Life sciences, 102(2), 98-104 (2014-03-25)
Cardiac hypertrophy is elicited by endothelin (ET)-1 as well as other neurohumoral factors, hemodynamic overload, and oxidative stress; HMG-CoA reductase inhibitors (statins) were shown to inhibit cardiac hypertrophy partly via the anti-oxidative stress. One of their common intracellular pathways is
Lee-Won Chong et al.
BMC gastroenterology, 15, 22-22 (2015-04-19)
Non-alcoholic steatohepatitis (NASH) is associated with hepatic fibrogenesis. Despite well-known cholesterol-lowering action of statins, their mechanisms against NASH-mediated fibrogenesis remain unclear. This study aimed at investigating the in vitro and in vivo anti-fibrotic properties of fluvastatin (Flu). Palmitate (PA)-induced changes
Hongjun Zhu et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 34(5), 539-543 (2014-11-25)
To observe the effect of Liandouqingmai recipe (Chinese herbal medicine compound preparation) on the quality of life (QOL) and inflammatory reaction of patients with coronary heart disease (CHD). A total of 101 CHD patients were randomized into two groups: treatment
Hege Pihlstrøm et al.
Transplantation, 98(11), 1219-1225 (2014-07-08)
Elevated symmetric dimethylarginine (SDMA) has been shown to predict cardiovascular events and all cause mortality in diverse populations. The potential role of SDMA as a risk marker in renal transplant recipients (RTR) has not been investigated. We analyzed SDMA in
Masaki Watanabe et al.
Drug metabolism and pharmacokinetics, 30(2), 198-204 (2015-05-21)
Since drug-drug interaction (DDI) can affect organic anion-transporting polypeptide (OATP) and cause clinical events, prediction of such DDI is important in early clinical development. Although statins are useful probes for OATP-mediated DDI, endogenous probes would be more practical for predicting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service