Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

236603

Sigma-Aldrich

C-Reactive Protein, Human Serum, High Purity

C-Reactive Protein, Human Serum, High Purity, CAS 9007-41-4, is a major acute-phase plasma protein. Useful as a calibration standard for assays of CRP and as an antigen for production of antisera.

Synonym(s):

C-Reactive Protein, Human Serum, High Purity, CRP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥99% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

M.W. 23,000 (monomer); 115,000 (pentamer).
Native C-reactive protein highly purified from human serum. CRP is a major acute-phase plasma protein that is dramatically elevated in patients with acute-phase conditions. Composed of five identical noncovalently linked subunits. Exhibits Ca2+-dependent binding to a variety of substrates. Important in diagnosis of rheumatoid arthritis. Suitable for immunological studies. In purified states and in plasma the monomeric form (M.W. ~21 kDa) aggregates as a cyclic pentamer. Intended for use as a calibration standard for assays of CRP, for structural and functional studies, and as an antigen for production of antisera.Calibrated by electroimmunoassay against WHO reference standard or equivalent.
No reaction in ODD and Laurell electroimmunoassay against polyvalent antiserum to normal human serum, antiserum to pooled effusion fluids and monospecific antiserum to human IgG.

Warning

Toxicity: Standard Handling (A)

Physical form

In 140 mM NaCl, 10 mM Tris, 2 mM CaCl₂, 0.1% NaN₃, pH 8.0.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Paffen, E., et al. 2004. Arterioscler. Thromb. Vasc. Biol.24, 975.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dobrin Nedelkov
Analytical chemistry, 79(15), 5987-5990 (2007-06-26)
Protein microarrays are the format of choice for high-throughput, high-content protein interaction analysis. In most of the array formats, reporter molecules are used in multistep detection of the protein interactions. Among the few existing label-free detection approaches, surface plasmon resonance
Elaine Paffen et al.
Arteriosclerosis, thrombosis, and vascular biology, 24(5), 975-981 (2004-03-27)
It is generally assumed that C-reactive protein (CRP) induces synthesis of tissue factor (TF) in monocytic cells, thereby potentially initiating intravascular blood coagulation. We aimed to elucidate the mechanism of CRP-induced TF expression in monocytes and monocyte-derived macrophages (MDMs) in
M Hundt et al.
European journal of immunology, 31(12), 3475-3483 (2001-12-18)
The classic acute-phase reactant C-reactive protein (CRP) plays an important role in innate immunity. Specific CRP receptors have been described on white blood cells and were further characterized as Fcgamma receptors I and II. Here, we used biotinylated, highly purified
B D Salkeld et al.
Neuroimmunomodulation, 9(6), 319-325 (2002-06-05)
This study examined the hypothesis that oral contraceptives (OC) influence the production of thermoregulatory cytokines, i.e. interleukin-6 (IL-6), soluble IL-6 receptor (sIL-6R), soluble glycoprotein 130 (s-gp130) and leptin, and that OC-induced changes in oral temperature (T(oral)) are associated with changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service