Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1617

Sigma-Aldrich

C Reactive Protein human

recombinant, expressed in E. coli, liquid

Synonym(s):

CRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

form

liquid

mol wt

23 kDa

bp

~100 °C

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... CRP(1401)

General description

C-reactive protein (CRP), a homopentameric acute-phase inflammatory protein, is produced mainly in liver hepatocytes. This highly conserved plasma protein is also produced by smooth muscle cells, macrophages, endothelial cells, lymphocytes, and adipocytes. It belongs to the pentraxin family.

Application

C Reactive Protein human has been used:
  • as a component in differentiation medium to study the effects of C-reactive protein (CRP) on the size of myotubes and myotube mixed protein synthesis (MPS)
  • to study the influence of serum CRP level from elderly on endothelial cell proliferation in human umbilical vein endothelial cells (HUVEC)
  • to study the roles and connections of nicotine, monocytic interleukin 6 (IL-6), α7 nicotinic acetylcholine receptor (α7-nAChR), and CRP in the development of coronary artery spasm (CAS)

Biochem/physiol Actions

C-reactive protein (CRP) is a protein that could precipitate the C-polysaccharide of pneumococcal cell walls. CRP is widely used as a clinical marker of the state of inflammation, since its production by hepatocytes increases during the acute phase of the inflammatory response.
C-reactive protein (CRP) plays a key role in the complement pathway, apoptosis, phagocytosis, nitric oxide (NO) release, and the production of interleukin-6 and tumor necrosis factor-α. It participates in the uptake of low-density lipoprotein in macrophages. CRP levels are elevated in people suffering from appendicitis, cholecystitis, pancreatitis, and meningitis. It plays a vital role in the pathophysiology of cardiovascular disease. Hence it is used as a marker of infection and cardiovascular events.

Analysis Note

A highly purified protein and made by a fermentation process using genetically modified E. coli.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P P Galanis et al.
Talanta, 238(Pt 2), 123056-123056 (2021-11-22)
We report the use of a laser-based fabrication process in the creation of paper-based flow-through filters that when combined with a traditional lateral flow immunoassay provide an alternative pathway for the detection of a pre-determined analyte over a wide concentration
Ming-Yow Hung et al.
Free radical biology & medicine, 120, 266-276 (2018-04-03)
Smoking and high-sensitivity C-reactive protein (hs-CRP) are risk factors for coronary artery spasm (CAS), which is characterized by the increased interleukin-6 (IL-6) level and monocyte counts; however, limited data are available regarding the role of cigarette-embedded nicotine in the modulation
Adrian Hosford-Donovan et al.
Maturitas, 89, 52-57 (2016-05-18)
It is hypothesized that chronic systemic inflammation contributes to the age-related decline in cardiovascular function. The aim of the present study was to combine an assessment of the relationship between the serum level of C-reactive protein (CRP) and systolic and
Ajay Kumar Yagati et al.
Bioelectrochemistry (Amsterdam, Netherlands), 128, 165-174 (2019-04-21)
Most clinical tests for biomarker detection require the support of a laboratory, and the results are usually slow, less sensitive, and lack the possibility for Point-of-Care (PoC) testing. Further, with the increasing demand for sensitive, portable, rapid, and low-cost devices
Britta Wåhlin-Larsson et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(1), 267-278 (2017-11-14)
Mechanisms underlying the relationship between systemic inflammation and age-related decline in muscle mass are poorly defined. The purpose of this work was to investigate the relationship between the systemic inflammatory marker CRP and muscle mass in elderly and to identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service