Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219374

Millipore

Cetyltrimethylammonium Bromide, Molecular Biology Grade

Cationic detergent suitable for the isolation of high molecular weight DNA in plants and other organisms.

Synonym(s):

Cetyltrimethylammonium Bromide, Molecular Biology Grade, Cetrimide, CTAB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H42BrN
CAS Number:
Molecular Weight:
364.45
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21

grade

Molecular Biology

Quality Level

description

Merck USA index - 14, 2025

Assay

≥98% (TLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

170

color

white

cation traces

heavy metals (as Pb): ≤0.001%

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Br-].[N+](CCCCCCCCCCCCCCCC)(C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

General description

Cationic detergent suitable for the isolation of high molecular weight DNA in plants and other organisms.

Application


  • Optimization of a high-throughput CTAB-based protocol for the extraction of qPCR-grade DNA from rumen fluid, plant and bacterial pure cultures.: This study details the optimization of a high-throughput protocol using Cetyltrimethylammonium Bromide (CTAB) for extracting high-quality DNA suitable for quantitative PCR (qPCR) from various biological samples. The optimized protocol is significant for molecular biology applications as it ensures efficient and reliable DNA extraction, which is crucial for downstream genetic analyses. (Minas et al., 2011).


Warning

Toxicity: Harmful (C)

Preparation Note

Heating to 30-35°C may be required for complete solubilization.

Reconstitution

Following reconstitution store at room temprature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

471.2 °F - closed cup

Flash Point(C)

244 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service