Skip to Content
MilliporeSigma

Skip To

648314

Tris Buffer, 1.0 M, pH 8.0, Molecular Biology Grade

1.0 M liquid, pH 7.0-9.0, Molecular Biology Grade

1 of 1 reviewers took part in a promotion

Synonym(s):

Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
UNSPSC Code:
12161705
NACRES:
NA.25
MDL number:
Beilstein/REAXYS Number:
741883
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tris Buffer, 1.0 M, pH 8.0, Molecular Biology Grade, Sterile solution. Suitable for DNA and RNA applications. Tris buffer is useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.

grade

Molecular Biology

Quality Segment

description

RTECS - TY2900000

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

foreign activity

DNases, RNases, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

General description

Sterile solution. Suitable for DNA and RNA applications. Useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.

Application


  • Physicochemical cell disruption of Bacillus sp. for recovery of polyhydroxyalkanoates: future bioplastic for sustainability.: This research used Tris buffer, pH 8.0, for cell lysis and extraction processes, contributing to sustainable bioplastic production through efficient recovery of polyhydroxyalkanoates (Bhat et al., 2024).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

1–5 of 5 Questions  
  1. Dear Sir,  Are the compositions of  this product the same with T6664 Tris buffer(powder form)?

    1 answer
    1. Product 648314, a discontinued item, is a 1.0 M Tris buffer solution, pH 8.0, molecular biology grade, supplied as a sterile liquid and based on Tris/Tris base. Product T6664 is Tris Buffered Saline (TBS) powder; when one pouch is dissolved in 1 L water, the result is 0.05 M Tris buffered saline containing NaCl 0.138 M and KCl 0.0027 M. Therefore, this product contains saline components and is more diluted in Tris after reconstitution, whereas this product is a 1.0 M Tris buffer solution, so they are not compositionally equivalent.

      Helpful?

  2. How is this product sterilized?

    1 answer
    1. This product is sterilized by filtering the solution through a 0.2 micron filter.

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  5. Is Tris Buffer, 1.0 M, pH 8.0 the same as 1XTE?

    1 answer
    1. A 1 M Tris buffer solution is not equivalent to a 1 X Tris-EDTA (TE) buffer preparation. The composition of 1X TE buffer is 10mM TRIS-HCl and 1mM EDTA. Please be aware there are Tris-EDTA buffer solutions offered in the product portfolio. These options that are pH 8.0 are products 93283 and 8890. Please see the corresponding product information page for both of these items at the links below:
      https://www.sigmaaldrich.com/product/sial/93283
      https://www.sigmaaldrich.com/product/mm/8890op

      Helpful?

Reviews

1 of 1 reviewers took part in a promotion

Active Filters

    • Review 1
    • Votes 0
    5 out of 5 stars.

    Fast delivery!

    I have had 0 issues since switching brands to Sigma and receive it faster than I did from the previous supplier

    Helpful?