Skip to Content
MilliporeSigma
All Photos(1)

Documents

218755

Sigma-Aldrich

Caspase-4 Inhibitor I

The Caspase-4 Inhibitor I, also referenced under CAS 402832-01-3, controls the biological activity of Caspase-4. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-4 Inhibitor I, Ac-LEVD-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36N4O9
CAS Number:
Molecular Weight:
500.54
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A reversible caspase-4 (ICH-2) inhibitor.
A reversible caspase-4 inhibitor.

Biochem/physiol Actions

Cell permeable: no
Primary Target
caspase-4
Product does not compete with ATP.
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Leu-Glu-Val-Asp-CHO

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Talanian, R.V., et al. 1997. J. Biol. Chem. 272, 9677.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahara L Chaudhry et al.
Brain sciences, 12(4) (2022-04-22)
The outbreak of COVID-19 caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) signifies a serious worldwide concern to public health. Both transcriptome and proteome of SARS-CoV-2-infected cells synergize the progression of infection in host, which may exacerbate symptoms and/or
Jian Zhang et al.
Science immunology, 7(68), eabk2092-eabk2092 (2022-02-05)
Goblet cells and their main secretory product, mucus, play crucial roles in orchestrating the colonic host-microbe interactions that help maintain gut homeostasis. However, the precise intracellular machinery underlying this goblet cell-induced mucus secretion remains poorly understood. Gasdermin D (GSDMD) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service