Skip to Content
MilliporeSigma
All Photos(1)

Documents

218753

Sigma-Aldrich

Caspase-5 Inhibitor I

Single spot by TLC, solid, Caspase-5 inhibitor, Calbiochem®

Synonym(s):

Caspase-5 Inhibitor I, Z-WE(OMe)HD(OMe)-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H42FN7O10
Molecular Weight:
763.77
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Caspase-5 Inhibitor I, Z-WEHD-FMK is a cell-permeable, potent, irreversible caspase-5 inhibitor, that also inhibits the activity of Caspase 1, 4, and 8.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable and irreversible inhibitor of caspase-5 (ICErel-III, TY), as well as caspase-1 and caspase-4.
A potent, cell-permeable, and irreversible inhibitor of caspase-5. Strongly inhibits caspase-1. Also inhibits caspase-4 and caspase-8.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-1, caspase-4, caspase-5, caspase-8
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Trp-Glu(OMe)-His-Asp(OMe)-CH₂F*

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 4 months at 4°C.

Analysis Note

single spot by TLC

Other Notes

Garcia-Calvo, M., et al. 1998. J. Biol. Chem. 273, 32608.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Rano, T., et al. 1997. Chem. Biol. 4, 149.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service