Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203695

Sigma-Aldrich

bpV(phen)

A potent protein phosphotyrosine phosphatase inhibitor and insulin receptor kinase (IRK) activator.

Synonym(s):

bpV(phen), Potassium Bisperoxo(1,10-phenanthroline)oxovanadate (V), PTEN Inhibitor I, PTP Inhibitor VIII

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K[VO(O2)2C12H8N2]
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥99% (51V-NMR)

form

solid

potency

38 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C12H8N2.K.2H2O2.O.V/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;2*1-2;;/h1-8H;;2*1-2H;;/q;+1;;;;

InChI key

MQPXOTNOFVCXDA-UHFFFAOYSA-N

General description

A potent insulin receptor kinase (IRK) activator and protein phosphotyrosine phosphatase inhibitor. Excellent insulin mimetic for in vitro and in vivo use (15 µg/kg). Inhibits in situ dephosphorylation of autophosphorylated insulin receptors with greater than 1000-fold potency over sodium orthovanadate. Arrests proliferation of neuroblastoma NB 41 and glioma C6 cells at the G2/M transition of the cell cycle. Also reported to potently inhibit PTEN (IC50 = 38 nM). The water content varies from lot to lot and is supplied on the label along with the lot-specific molecular weight.
A potent protein phosphotyrosine phosphatase inhibitor and insulin receptor kinase (IRK) activator. Excellent insulin mimetic at 15 µg/kg in vitro and in vivo for use. Inhibits the in situ dephosphorylation of autophosphorylated insulin receptors with over 1000-fold greater potency than sodium orthovanadate. Arrests proliferation of neuroblastoma NB 41 and glioma C6 cells at the G2/M transition of the cell cycle. Also reported to potently inhibit PTEN (IC50 = 38 nM).
The water content varies from lot to lot and is supplied on the label along with the lot-specific molecular weight.

Biochem/physiol Actions

Cell permeable: no
Primary Target
PTEN
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Schmid, A.C., et al. 2004. FEBS Lett.566, 35.
Muzyamba, M.C., et al. 1999. J. Physiol. 517, 421.
Drake, P.G., et al. 1996. Endocrinology137, 4960.
Bevan, A.P., et al. 1995. Am. J. Physiol.268, E60.
Bevan, A.P., et al. 1995. J. Biol. Chem. 270, 10784.
Yale, J.F., et al. 1995. Diabetes 44, 1274.
Posner, B.I., et al. 1994. J. Biol. Chem. 269, 4596.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameer S Chopra et al.
Cell systems, 10(1), 66-81 (2019-12-10)
Frequent mutation of PI3K/AKT/mTOR signaling pathway genes in human cancers has stimulated large investments in targeted drugs but clinical successes are rare. As a result, many cancers with high PI3K pathway activity, such as triple-negative breast cancer (TNBC), are treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service