Skip to Content
MilliporeSigma

12-151

PKA Inhibitor Peptide

lyophilized powder, for Neuroscience applications

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

PKA Inhibitor peptide, The PKA Inhibitor peptide controls the biological activity of PKA. This small molecule/inhibitor is primarily used for Neuroscience applications.

Quality Level

form

lyophilized powder

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Biochem/physiol Actions

Protein Target: PKA
Target Sub-Family: AGC

Physical form

Lyophilized powder (from H2O)

Preparation Note

Lyophilized: Stable for 2 years at -20°C . Rehydrated: Stable for 1 year at -20°C.

Analysis Note

Routinely evaluated by inhibition of PKA catalytic subunit (14-114).

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Heterogeneity in beta-adrenergic receptor kinase expression in the lung accounts for cell-specific desensitization of the beta2-adrenergic receptor
McGraw, D. W. and Liggett, S. B.
The Journal of Biological Chemistry, 272, 7338-7344 (1997)
Y I Lee et al.
The Journal of biological chemistry, 276(20), 16969-16977 (2001-03-30)
The hepatitis B virus-X (HBx) protein is known as a multifunctional protein that not only coactivates transcription of viral and cellular genes but coordinates the balance between proliferation and programmed cell death, by inducing or blocking apoptosis. In this study
H L Lai et al.
The Journal of biological chemistry, 272(8), 4970-4977 (1997-02-21)
We have previously reported that phosphorylation of adenylyl cyclase type VI (AC6) may result in the suppression of adenylyl cyclase activity during desensitization of the A2a-adenosine receptor-mediated cAMP response (A2a desensitization) in rat pheochromocytoma PC12 cells. In the present study



Global Trade Item Number

SKUGTIN
12-15104053252333507