Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.94512

Supelco

Ethyl acetate

EMPLURA®

Synonym(s):

Ethyl acetate, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12352108

grade

for analytical purposes
for cleaning
for extraction
for preparative purposes
for synthesis

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

product line

EMPLURA®

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical

format

neat

storage temp.

20-25°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service