Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

1.04735

Supelco

Water

tested for EPA533 and EPA537.1 PFAS Methods LiChrosolv®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
41104213

description

suitable for EPA 533
suitable for EPA 537.1

Quality Level

Assay

≥99.9% (GC)

form

liquid

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

suitability

suitable for UHPLC-MS

SMILES string

O

application(s)

PFAS testing

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrosolv ultra-high purity solvents for PFAS testing are specially designed for this high demanding application. The products will help to reduce background interference and will not contain any of the PFAS compounds above the limit values defined by the EPA 533 and EPA 537.1 methods. This allows reliable and effective testing to facilitate identification and quantitation of PFAS analytes and eliminates the risk of false positive or false negative results in LC-MS/MS analysis.
PFAS tested solvents are used for UHPLC-MS and LC-MS instrumental analysis.

Application

PFAS tested solvents are qualified for PFAS analysis according to drinking water testing methods EPA 533 and EPA 537.1.

PFAS testing features:
- QC Batch tested 29 PFAS analytes according to EPA 533 (25 analytes) and EPA 537.1 (18 analytes)
- Lot to lot consistency: Multiple batches validated by a third-party accredited testing laboratory and internal laboratories

Features and Benefits

- ESI/APCI (+) < 2 ppb; ESI/APCI (-) < 10 ppb
- Lowest impurity profile for Interference free baseline
- Borosilicate glass bottles minimize contamination with metal ions
- Lowest levels of trace metal impurities: for minimized metal ion adduct formation <5ppb
- Microfiltration through 0.2 μm filter

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sensitive PFAS testing can be performed with ultrapure water from a Milli-Q® IQ 7 system, which eliminates detectable PFAS for reliable LC-MS/MS results. See the science, see the proof.

Sensitive PFAS testing can be performed with ultrapure water from a Milli-Q® IQ 7 system, which eliminates detectable PFAS for reliable LC-MS/MS results. See the science, see the proof.

Sensitive PFAS testing can be performed with ultrapure water from a Milli-Q® IQ 7 system, which eliminates detectable PFAS for reliable LC-MS/MS results. See the science, see the proof.

Sensitive PFAS testing can be performed with ultrapure water from a Milli-Q® IQ 7 system, which eliminates detectable PFAS for reliable LC-MS/MS results. See the science, see the proof.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service