Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.94500

Supelco

Acetone

for preparative purposes, EMPLURA®

Synonym(s):

Acetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12352115
grade:
for extraction
for synthesis
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

grade

for extraction
for synthesis

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

description

for analytical purposes

product line

EMPLURA®

form

liquid

expl. lim.

13.2 %

dilution

(low-cost alternative, for basic applications)

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical

format

neat

storage temp.

room temp

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Packaging

1.94500.0521: 500ML, Minimum order quantity: 20
1.94500.2521: 2.5L, Minimum order quantity: 4
1.94500.9027: 25 L Minimum Order Quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service