Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93024

Supelco

Hydriodic acid 55%

for analysis EMPARTA® ACS

Synonym(s):

Hydriodic acid, Hydriotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HI
CAS Number:
Molecular Weight:
127.91
MDL number:
UNSPSC Code:
12352106

grade

ACS reagent

bp

127 °C (lit.)

storage temp.

room temp

SMILES string

I

InChI

1S/HI/h1H

InChI key

XMBWDFGMSWQBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93024.0251: 250 mL, Minimum order quantity: 12

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service