Skip to Content
MilliporeSigma

1.00344

Hydroiodic acid

57%, for analysis EMSURE®

Synonym(s):

Hydrogen iodide solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352301
NACRES:
NA.21
Assay:
≥57% (acidimetric)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hydroiodic acid 57%, for analysis EMSURE®

Quality Segment

product line

EMSURE®

assay

≥57% (acidimetric)

form

liquid

impurities

≤0.1 ppm Ba (Barium), ≤0.1 ppm Be (Beryllium), ≤0.1 ppm Bi (Bismuth), ≤0.1 ppm Cd (Cadmium), ≤0.1 ppm Co (Cobalt), ≤0.1 ppm Cr (Chromium), ≤0.1 ppm Cu (Copper), ≤0.1 ppm Ge (Germanium), ≤0.1 ppm Li (Lithium), ≤0.1 ppm Mn (Manganese), ≤0.1 ppm Mo (Molybdenum), ≤0.1 ppm Ni (Nickel), ≤0.1 ppm Pb (Lead), ≤0.1 ppm Sr (Strontium), ≤0.1 ppm Ti (Titanium), ≤0.1 ppm Tl (Thallium), ≤0.1 ppm V (Vanadium), ≤0.1 ppm Zr (Zirconium), ≤0.5 ppm Zn (Zinc), ≤1 ppm Al (Aluminium), ≤1 ppm As (Arsenic), ≤1 ppm Ca (Calcium), ≤1 ppm K (Potassium), ≤1 ppm Na (Sodium), ≤10 ppm Phosphorous compounds (as PO4), ≤100 ppm Chloride and Bromide (as Cl), ≤2 ppm Fe (Iron), ≤20 ppm Sulfate (SO4), ≤5 ppm Mg (Magnesium)

ign. residue

≤50 ppm (as sulfate)

pH

1 (20 °C, 13 g/L in H2O)

bp

127 °C/1013 hPa

mp

-50 °C (External MSDS)

density

1.70 g/cm3 at 20 °C

storage temp.

2-30°C

Application


  • Spontaneous anion-exchange synthesis of optically active mixed-valence Cs(2)Au(2)I(6) perovskites from layered CsAuCl(4) perovskites: This research highlights the use of 57% hydroiodic acid in the synthesis of advanced perovskite materials, showcasing its effectiveness in facilitating high-purity chemical transformations for material science applications. The study provides valuable insights into the generation of optically active materials for potential use in electronic and optical devices (Bhawna et al., 2021).

  • Analytical challenges in the quantitative determination of 2H/1H ratios of methyl iodide: This study employs 57% hydroiodic acid in the preparation of samples for isotopic analysis in mass spectrometry, demonstrating its crucial role in environmental testing and analytical chemistry. The research addresses significant challenges in measuring isotopic compositions, which are essential for tracing chemical sources and pathways in environmental science (Feakins et al., 2013).

Analysis Note

Assay (acidimetric): ≥ 57 %
Chloride and Bromide (as Cl): ≤ 100 ppm
Phosphorous compounds (as PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 20 ppm
Al (Aluminium): ≤ 1 ppm
As (Arsenic): ≤ 1 ppm
Ba (Barium): ≤ 0.1 ppm
Be (Beryllium): ≤ 0.1 ppm
Bi (Bismuth): ≤ 0.1 ppm
Ca (Calcium): ≤ 1 ppm
Cd (Cadmium): ≤ 0.1 ppm
Co (Cobalt): ≤ 0.1 ppm
Cr (Chromium): ≤ 0.1 ppm
Cu (Copper): ≤ 0.1 ppm
Fe (Iron): ≤ 2 ppm
Ge (Germanium): ≤ 0.1 ppm
K (Potassium): ≤ 1 ppm
Li (Lithium): ≤ 0.1 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 0.1 ppm
Mo (Molybdenum): ≤ 0.1 ppm
Na (Sodium): ≤ 1 ppm
Ni (Nickel): ≤ 0.1 ppm
Pb (Lead): ≤ 0.1 ppm
Sr (Strontium): ≤ 0.1 ppm
Ti (Titanium): ≤ 0.1 ppm
Tl (Thallium): ≤ 0.1 ppm
V (Vanadium): ≤ 0.1 ppm
Zn (Zinc): ≤ 0.5 ppm
Zr (Zirconium): ≤ 0.1 ppm
Alkoxygroups: passes test
Residue on ignition (as sulfate): ≤ 50 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Hydroiodic acid 57%


Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point (°F)

does not flash

Flash Point (°C)

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library