Skip to Content
MilliporeSigma

1.00341

Hydroiodic acid

57%, EMPLURA®

Synonym(s):

Hydrogen iodide solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352301
NACRES:
NA.21
Assay:
≥57% (acidimetric)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hydroiodic acid 57%, EMPLURA®

Quality Segment

product line

EMPLURA®

assay

≥57% (acidimetric)

form

liquid

impurities

≤0.0005% Fe (Iron), ≤0.001% Heavy metals (as Pb), ≤0.005% Sulfate (SO4), ≤0.03% Chloride and Bromide (as Cl), ≤0.06% Non volatile matter

pH

1 (20 °C, 13 g/L in H2O)

bp

127 °C/1013 hPa

mp

-50 °C (External MSDS)

density

1.70 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): ≥ 57 %
Chloride and Bromide (as Cl): ≤ 0.03 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Non volatile matter: ≤ 0.06 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Hydroiodic acid 57%


Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point (°F)

does not flash

Flash Point (°C)

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library