Skip to Content
MilliporeSigma
All Photos(2)

Documents

16-316

Sigma-Aldrich

Anti-Phosphotyrosine Antibody, 4G10® Platinum, HRP Conjugate

Upstate®, from mouse, peroxidase conjugate

Synonym(s):

phosphotyrosine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody product type

primary antibodies

clone

4G10®, monoclonal
PY20, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

Gene Information

human ... PID1(55022)

General description

The development of the anti-phosphotyrosine, clone 4G10 in 1989 was a monumental discovery for researchers. 4G10 was the first and is the best single monoclonal antibody for the detection tyrosine phosphorylation. 4G10 is well known for its sensitivity and its ability to detect multiple tyrosine phosphorylations on numerous substrates. It has been validated by thousands of scientific and medical researchers in virtually every application and tyrosine target over the past 2 decades. To improve on something that hundred have tried and no one has succeeded, we pooled 4G10 with the next most highly regarded anti-phosphotyrosine, clone PY20 to make 4G10 Platinum. PY20 itself is a very poor substitute for 4G10, but its additive effect allow for a greater level of detection on more substrates that even 4G10 alone.

Application

Detect Phosphotyrosine with Anti-Phosphotyrosine Antibody, 4G10 Platinum, HRP Conjugate (Mouse Monoclonal Antibody), that has been shown to work in WB, ELISA.

Quality

Routinely evaluated by western blot.

Target description

Varies

Physical form

100 μL of a proprietary mixture of protein G purified mouse monoclonal 4G10 (IgG2bƙ) HRP conjugate and protein A purified mouse monoclonal PY20, HRP conjugate (IgG2b) in PBS containing 0.05% Kathon and HRP stabilizing compounds. Sufficient for a minumum of 10 immunoblots, if used at the optimum concentration of 1:2000 – 1:4000 in 10 mL blocking buffer. Liquid.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irmgard Hofmann et al.
The Journal of pharmacology and experimental therapeutics (2022-10-15)
VEGF and angiopoietin-2 (ANG2) have complementary roles in angiogenesis and promote an immunosuppressive tumor microenvironment. It is anticipated that combination of VEGF and ANG2 blockade could provide superior activity to blockade of either pathway alone, and that addition of VEGF/ANG2
T Wang et al.
Nihon Rai Gakkai zasshi, 60(3-4), 146-151 (1991-07-01)
A comparative study on the usefulness of Kawatsu's silver impregnating staining method compared with S-100 protein ABC technique for the differential diagnosis of tuberculoid leprosy was carried out. The results of neurohistological examination obtained from both methods were almost the
Ondřej Ballek et al.
Immunology letters, 142(1-2), 64-74 (2012-01-28)
Lck is the principal signal-generating tyrosine kinase of the T cell activation mechanism. We have previously demonstrated that induced Lck activation outside of lipid rafts (LR) results in the rapid translocation of a fraction of Lck to LR. While this
Guillermina M Goñi et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(31), E3177-E3186 (2014-07-23)
Focal adhesion kinase (FAK) is a nonreceptor tyrosine kinase (NRTK) with key roles in integrating growth and cell matrix adhesion signals, and FAK is a major driver of invasion and metastasis in cancer. Cell adhesion via integrin receptors is well
Kogulan Yoganathan et al.
Frontiers in immunology, 13, 848577-848577 (2022-08-23)
The E protein transcription factors E2A and HEB are critical for many developmental processes, including T cell development. We have shown that the Tcf12 locus gives rise to two distinct HEB proteins, with alternative (HEBAlt) and canonical (HEBCan) N-terminal domains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service