Skip to Content
MilliporeSigma

16-455

Anti-Phosphoserine Antibody, clone 4A4, HRP Conjugate

clone 4A4, Upstate®, from mouse

Synonym(s):

4A4, anti-phosphoserine, phosphoserine antibody, pan-phosphoserine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
peroxidase conjugate
Clone:
4A4, monoclonal
Application:
WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

peroxidase conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

4A4, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

phosphorylation (pSer)

General description

The identification of protein phosphorylation as a regulatory mechanism originated from studies by Fischer and Krebs in the mid 1950s that later earned them the 1992 Nobel prize. It is the major mechanism for the regulation of diverse cellular processes including cell division, protein synthesis, transcriptional regulation and neurotrans-mission. The steady state phosphorylation of any given substrate is governed by the opposing activities of kinases and phosphatases. It is now believed that a third of all eukaryotic cellular proteins are phosphorylated and that over 95% of all phosphorylation events occur on serine and threonine residues.
Varies

Immunogen

Epitope: phosphoserine
Phosphoserine conjugated to KLH.

Application

Anti-Phosphoserine Antibody, clone 4A4, HRP Conjugate is an antibody against Phosphoserine for use in WB.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Biochem/physiol Actions

Recognizes proteins containing phosphoserine residues, regardless of origin.

Physical form

100 μL of protein G purified mouse IgG1 in PBS containing preservative and HRP stabilizing compounds. Sufficient for a minimum of 10 immunoblots, if used at the optimum concentration of 1:4000 in 10 mL blocking buffer. Liquid.
Protein G Purified

Preparation Note

6 months at 2-8°C from date of shipment.
NOTE: DO NOT FREEZE. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Routinely evaluated by western blot on RIPA lysates from insulin-treated human HEK293 cells.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library