MilliporeSigma
  • The use of improved silver impregnating staining method in the differential diagnosis of tuberculoid leprosy.
The use of improved silver impregnating staining method in the differential diagnosis of tuberculoid leprosy.
Nihon Rai Gakkai zasshi (1991-07-01)
T Wang, K I Butt, Y Maeda, K Kawatsu, S Izumi
ABSTRACT
A comparative study on the usefulness of Kawatsu's silver impregnating staining method compared with S-100 protein ABC technique for the differential diagnosis of tuberculoid leprosy was carried out. The results of neurohistological examination obtained from both methods were almost the same. It can be said that Kawatsu's method is useful and cost-effective, and thus suitable for practical use in developing countries.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, 4G10® Platinum, HRP Conjugate, Upstate®, from mouse, peroxidase conjugate