Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-738-M

Sigma-Aldrich

Pim-3 Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, human Pim-3, for use in Enzyme Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 39.6 kDa

purified by

(Ni2+/NTA-agarose)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

accession no.

 (FUNCTION: SwissProt: Q86V86 # May be involved in cell cycle progression and anti- apoptosis process. Implicated in proliferation of human hepatoma cell lines. )
NM_001001852.2

NCBI accession no.

UniProt accession no.

relevant disease(s)

cardiovascular diseases; metabolic disorders

General description

N-terminal 6His-tagged, recombinant, human Pim-3

Biochem/physiol Actions

Protein Target: Pim-3

Packaging

Also available in 10 μg size and in bulk

Quality

routinely evaluated by phosphorylation of 250μM GGMEDIYFEFMGGKKK

Target description

Pim-3

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service