Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475815-M

Sigma-Aldrich

Microcystin-LR, Microcystis aeruginosa

Cyclic heptapeptide toxin isolated from the freshwater cyanobacteria Microcystis aeruginosa.

Synonym(s):

Microcystin-LR, Microcystis aeruginosa, MCYST-LR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H74N10O12
CAS Number:
Molecular Weight:
995.17
MDL number:
UNSPSC Code:
12352200

Quality Level

description

RTECS - GT2810000

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

pale yellow

solubility

methanol: 2 mg/mL
DMSO: 2.5 mg/mL

storage temp.

−20°C

SMILES string

N1[C@H]([C@@H](C(=O)N[C@H](CCC(=O)N(C(=C)C(=O)N[C@@H](C(=O)N[C@H](C(=O)N[C@H]([C@@H](C(=O)N[C@H](C1=O)CCCN=C(N)N)C)C(=O)O)CC(C)C)C)C)C(=O)O)C)\C=C\C(=C\[C@@H]([C@@H](OC)Cc2ccccc2)C)\C

InChI

1S/C49H74N10O12/c1-26(2)23-37-46(66)58-40(48(69)70)30(6)42(62)55-35(17-14-22-52-49(50)51)45(65)54-34(19-18-27(3)24-28(4)38(71-10)25-33-15-12-11-13-16-33)29(5)41(61)56-36(47(67)68)20-21-39(60)59(9)32(8)44(64)53-31(7)43(63)57-37/h11-13,15-16,18-19,24,26,28-31,34-38,40H,8,14,17,20-23,25H2,1-7,9-10H3,(H,53,64)(H,54,65)(H,55,62)(H,56,61)(H,57,63)(H,58,66)(H,67,68)(H,69,70)(H4,50,51,52)/b19-18+,27-24+/t28-,29-,30-,31+,34-,35-,36+,37-,38-,40+/m0/s1

InChI key

ZYZCGGRZINLQBL-GWRQVWKTSA-N

General description

Cyclic heptapeptide toxin isolated from the freshwater cyanobacteria Microcystis aeruginosa. Potent inhibitor of protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A). Unlike Okadaic Acid (Cat. No. 495604), microcystin-LR is effective on both PP1 (IC50 = 1.7 nM) and PP2A (IC50 = 40 pM). Reacts with phosphatase in a two-step mechanism involving rapid binding and inactivation of the catalytic subunit followed by slower covalent interactions. Has no effect on protein kinases, making it useful for reducing the effect of contaminating phosphatase activities in protein kinase assays. A 2.5 mM (250 µg/100 µl) solution of Microcystin-LR, Microcystis aeruginosa (Cat. No. 475821) in DMSO is also available.
Cyclic heptapeptide toxin isolated from the freshwater cyanobacteria, Microcystis aeruginosa. Potent inhibitor of protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A). Unlike Okadaic Acid (Cat. No. 495604), microcystin-LR is effective on both PP1 (IC50 = 1.7 nM) and PP2A (IC50 = 40 pM). Has no effect on protein kinases, making it useful for reducing contaminating phosphatase activities in protein kinase assays.

Biochem/physiol Actions

Target IC50: 1.7 nM and 40 pM against PP1 and PP2A, respectively

Warning

Toxicity: Highly Toxic (H)

Preparation Note

Further dilute with aqueous buffer just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Unstable at > pH 7.7.

Other Notes

Craig, M., et al. 1996. Biochem. Cell Biol. 74, 569.
Nishiwaki, R., et al. 1994. Cancer Lett. 83, 283.
Amick, G.D., et al. 1992. Biochem. J.287, 1019.
Davidson, H.W., et al. 1992. J. Cell Biol.116, 1343.
Nishiwaki-Matsushima, R., et al. 1992. J. Cancer Res. Clin. Oncol. 118, 420.
Cohen, P., et al. 1990. Trends Biochem. Sci.15, 98.
MacKintosh, C., et al. 1990. FEBS Lett.264, 187.
Cohen, P., et al. 1989. FEBS Lett.250, 596.
Carmichael, W.W. 1988. in Handbook of Natural Toxins3: Marine Toxins and Venoms(Tu, A.T., Ed.) p. 121, Decker, New York.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service