Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46366

Millipore

Sabouraud Dextrose broth

bottle capacity 125 mL, bottle filling volume 100 mL, closure type, screw cap with septum, box of 10 bottles

Synonym(s):

Ready-to-use SDB, Sabouraud Liquid Medium, SDB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP 2.6.1
USP 62

Quality Level

description

For the cultivation and isolation of yeasts, molds, & dermatophytes; Suitable for sterility testing

sterility

sterile

form

liquid (ready-to-use)

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

box of 10 bottles

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

125 mL

bottle filling volume

100 mL

pH

5.6

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Broth (SDB) is a complex medium for cultivation and isolation of yeasts, molds, and dermatophytes. The high concentration of Dextrose and low pH promotes the growth of yeast and molds while inhibiting bacterial growth. The media also promotes the formation of spores (conidia and sporangia), as well as the formation of pigments of yeasts and molds.

Application

Sabouraud Dextrose Broth (SDB) is used for the cultivation and isolation of yeasts, molds, and dermatophytes. The media is suitable for the microbiological examination of non-sterile products and the determination of the fungistatic activity of pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Our media are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service