Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46073

Millipore

HEIMPLATE XLD Agar - LI EP+USP 30ml

Selective media for the isolation of Salmonella and Shigella in pharmaceuticals and clinical materials

Synonym(s):

Xylose Lysine Deoxycholate Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Agency

EP 2.6.12
USP 61

Quality Level

sterility

sterile

form

solid

feature

ready-to-use

technique(s)

microbiological culture: suitable

pH

7.4 ( in H2O)

application(s)

microbiology
pharmaceutical

storage temp.

20-25°C

suitability

Salmonella spp. (spp.)

General description

Xylose Lysine Deoxycholate agar (XLD agar) is a selective growth medium introduced by Taylor (1965) for the isolation of Salmonella and Shigella species (Taylor and schelbert, 1971). It relies on the use of selective inhibitory component sodium deoxycholate and three indicator systems, i.e. xylose, lactose and sucrose combined with phenol red. Degradation of xylose, lactose, and sucrose to acid causes phenol red to change its colour to yellow. Sodium thiosulfate and iron (III) salt react to form a precipitate of black iron sulfide in the colonies, indicating the production of hydrogen sulfide. Bacteria which decarboxylate lysine to cadaverine can be recognized by the appearance of a purple coloration around the colonies due to an increase in pH. These reactions can proceed simultaneously or successively. This causes the pH indicator to exhibit various shades of colour, or it may change its colour from yellow to red on prolonged incubation. The culture medium is weakly inhibitory to other microbes due to the presence of sodium deoxycholate.

Application

Xylose Lysine Deoxycholate (XLD) Agar - LI is a selective culture medium for isolation of Salmonella and Shigella in pharmaceuticals.

Packaging

90 mm settle plates with 30 ml filling volume (20 or 120 plates per box) for long incubation (LI)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of culture media for food and water microbiology
Curtis, GDW and Baird, Rosamund M and Corry, Janet EL et al.
Royal Society of Chemistry (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service