Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44257

Millipore

RCS Compressed Gas Adapter Touch

for use with RCS® High Flow Touch, for testing compressed gas

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

aluminum adapter

Agency

non-compendial

description

pressure head

product line

RCS®

feature

portable

packaging

pkg of 1 kit

application(s)

compressed gas monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

room temp

General description

RCS® Compressed Gas Adapter Touch is an additional accessory for use with RCS® High Flow Touch air sampler. The RCS® High Flow Touch is a portable air sampler for fast and easy monitoring of ambient air and compressed gases in controlled environments. The microbial air sampler is based on impaction by centrifugal force. The RCS® Compressed Gas Adapter Touch is autoclavable and is used for microbial monitoring of compressed gases. It is designed for an air inlet pressure of 1 bar.

Application

RCS® Compressed Gas Adapter Touch is an additional accessory with the RCS® microbial air sampler used for microbial monitoring of compressed gases. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service