Skip to Content
MilliporeSigma

1.44243

Hycon® Agar Strips SDX

Sabouraud Dextrose Agar, Chlortetracycline, Chloramphenicol, for yeasts, for molds, aseptically filled, single packed of 50 units, suitable for air monitoring

Synonym(s):

Hycon Ready-to-use Agar Strips, SDX, SDA, Agar Strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41113035
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

aseptically filled

product line

RCS®

form

plate (ready-to-use)

feature

individually wrapped

composition

Chloramphenicol , Chlortetracycline

packaging

single packed of 50 units

manufacturer/tradename

HYCON®

pH

6.9 ( in H2O)

application(s)

air monitoring, cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

2-25°C

suitability

molds, yeasts

General description

Agar Strips SDX are ready-to-use culture media for assessment of airborne yeasts and molds with RCS® Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. Sabouraud Dextrose Agar is filled in a flexible backing film. Sabouraud Dextrose Agar (SDX, SDA) is a complex medium for cultivation and isolation of a wide range of yeasts and molds. Dextrose supports the growth of fungi. The medium is supplemented with chloramphenicol and chlortetracycline to inhibit the growth of accompanying bacterial flora.

Application

Agar Strips are used for monitoring of ambient air and compressed gases with RCS® Air Samplers.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library