Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44152

Millipore

USB Adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

plastic adapter

Agency

non-compendial

description

For use with High Flow Touch
interface

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

shipped in

ambient

storage temp.

room temp

General description

USB Adapter is an additional accessory for use with RCS® High Flow microbial air sampler. The RCS® High Flow is a portable air sampler for fast and easy monitoring of ambient air and compressed gases in controlled environments. This microbial air sampler is based on impaction by centrifugal force and can also be used with a compressed gas adapter for additional monitoring of compressed gases. The USB Adapter is used for bridging the connection of the serial RS232 interface of RCS® instruments with the USB interface of a PC. The adaptor is also compatible for use with RCS® Instruments of the RCS® Isolator and RCS® Anemometer.

Application

USB Adapter is an additional accessory with the RCS® High Flow air sampler used for connecting the RCS® instruments with the USB interface of a PC. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service