MilliporeSigma
Heat shock protein 27 controls apoptosis by regulating Akt activation.
The Journal of biological chemistry (2003-05-13)
Madhavi J Rane, Yong Pan, Saurabh Singh, David W Powell, Rui Wu, Timothy Cummins, Qingdan Chen, Kenneth R McLeish, Jon B Klein
ABSTRACT
Activation of the serine-threonine kinase Akt by cytokines, chemokines, and bacterial products delays constitutive neutrophil apoptosis, resulting in a prolonged inflammatory response. We showed previously that Akt exists in a signaling complex with p38 MAPK, MAPK-activated protein kinase-2 (MAPKAPK-2), and heat shock protein-27 (Hsp27); and Hsp27 dissociates from the complex upon neutrophil activation. To better understand the regulation of this signaling module, the hypothesis that Akt phosphorylation of Hsp27 regulates its interaction with Akt was tested. The present study shows that Akt phosphorylated Hsp27 on Ser-82 in vitro and in intact cells, and phosphorylation of Hsp27 resulted in its dissociation from Akt. Additionally, the interaction between Hsp27 and Akt was necessary for activation of Akt in intact neutrophils. Constitutive neutrophil apoptosis was accelerated by sequestration of Hsp27 from Akt, and this enhanced rate of apoptosis was reversed by introduction of constitutively active recombinant Akt. Our results define a new mechanism by which Hsp27 regulates apoptosis, through control of Akt activity.
Sigma-Aldrich
MAPKAP Kinase 2 Protein, active, 10 µg, Active, for use in Kinase Assays.
Sigma-Aldrich
Akt1/PKBα Protein, active, 15 µg, Active, recombinant full-length human Akt1, containing N-terminal His6 tag, for use in IP-Kinase Assays & Kinase Assays.