MilliporeSigma
  • MAPKAP kinase-2; a novel protein kinase activated by mitogen-activated protein kinase.
MAPKAP kinase-2; a novel protein kinase activated by mitogen-activated protein kinase.
The EMBO journal (1992-11-01)
D Stokoe, D G Campbell, S Nakielny, H Hidaka, S J Leevers, C Marshall, P Cohen
ABSTRACT
A novel protein kinase, which was only active when phosphorylated by the mitogen-activated protein kinase (MAP kinase), has been purified 85,000-fold to homogeneity from rabbit skeletal muscle. This MAP kinase activated protein kinase, termed MAPKAP kinase-2, was distinguished from S6 kinase-II (MAPKAP kinase-1) by its response to inhibitors, lack of phosphorylation of S6 peptides and amino acid sequence. MAPKAP kinase-2 phosphorylated glycogen synthase at Ser7 and the equivalent serine (*) in the peptide KKPLNRTLS*VASLPGLamide whose sequence is similar to the N terminus of glycogen synthase. MAPKAP kinase-2 was resolved into two monomeric species of apparent molecular mass 60 and 53 kDa that had similar specific activities and substrate specificities. Peptide sequences of the 60 and 53 kDa species were identical, indicating that they are either closely related isoforms or derived from the same gene. MAP kinase activated the 60 and 53 kDa forms of MAPKAP kinase-2 by phosphorylating the first threonine residue in the sequence VPQTPLHTSR. Furthermore, Mono Q chromatography of extracts from rat phaeochromocytoma and skeletal muscle demonstrated that two MAP kinase isoforms (p42mapk and p44mapk) were the only enzymes in these cells that were capable of reactivating MAPKAP kinase-2. These results indicate that MAP kinase activates at least two distinct protein kinases, suggesting that it represents a point at which the growth factor-stimulated protein kinase cascade bifurcates.
Sigma-Aldrich
MAPKAP Kinase 2 Protein, active, 10 µg, Active, for use in Kinase Assays.
Sigma-Aldrich
MAPKAP Kinase 2 Protein, inactive, 50 g, Unactive, For use in Kinase Assays.
Sigma-Aldrich
MAP Kinase 2/Erk2 Protein, active, mouse, 10 µg, Active, N-terminal GST-tagged, recombinant MAP Kinase 2, expressed in E. coli cells, activated using MEK1 (Catalog #14-429). For use in Kinase Assays.