Skip to Content
MilliporeSigma

1.06573

di-Sodium hydrogen phosphate dodecahydrate

crystalline, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP

Synonym(s):

Sodium phosphate dibasic dodecahydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Na2HPO4 · 12H2O
CAS Number:
Molecular Weight:
358.14
UNSPSC Code:
12352302
EC Index Number:
231-448-7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, ChP, JP, Ph. Eur., USP

product line

EMPROVE® EXPERT

form

crystalline, solid

pH

9.0-9.3 (20 °C, 50 g/L in H2O)

solubility

218 g/L

density

1.52 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.12H2O/c;;1-5(2,3)4;;;;;;;;;;;;/h;;(H3,1,2,3,4);12*1H2/q2*+1;;;;;;;;;;;;;/p-2

InChI key

DGLRDKLJZLEJCY-UHFFFAOYSA-L

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

di-Sodium hydrogen phosphate dodecahydrate is typically used as a phosphate buffer system component for biomolecule downstream chromatography steps or final liquid formulation.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library