Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.25163

Millipore

Eshmuno® A Resin

RoboColumn Eshmuno® A, volume 0.6 mL, prepacked column

Synonym(s):

RoboColumn Eshmuno® A, 0.6ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

ligand

protein A

Quality Level

product line

Eshmuno®

form

prepacked column

manufacturer/tradename

Eshmuno®

parameter

≤8 bar pressure
~500 cm/hr max. flow rate

column diam. × L

5 mm × 30 mm

volume

0.6 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

capacity

~40-55 mg/mL, resin binding capacity((3-6 min RT))

application(s)

mAb

storage temp.

2-8°C

General description

5 x 30mm ESHMUNO®

Features and Benefits

Eshmuno® A chromatography resin enables users to:

  • Intensify processes resulting in High Binding Capacity, Superior Flow Capability, and Excellent HCP reduction.
  • Determine the products purity with a more consistent viral clearance, increased aggregate removal, reduced fab finding and low leached Protein A.

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service