Skip to Content
MilliporeSigma

1.20089

Millipore

Eshmuno® A Resin

Eshmuno® A, affinity

Synonym(s):

Eshmuno® A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

ligand

protein A

Quality Level

form

resin

specific analyte(s)

MAbs

manufacturer/tradename

Eshmuno®

parameter

>500 cm/hr flow rate
8 bar max. pressure

matrix active group

polymer, polyvinyl ether

mean particle size

50 μm

capacity

40-55 mg/mL binding capacity

transition temp

flash point 35 °C (calculated)

separation technique

affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Protein A affinity resin based on polyvinyl ether base matrix

Features and Benefits

Eshmuno® A chromatography resin provides users with:

  • An Intensified processes resulting in High Binding Capacity, Superior Flow Capability, and Excellent HCP reduction.
  • Products purity with a more consistent viral clearance, increased aggregate removal, reduced fab finding and low leached Protein A
  • High dynamic binding capacity at high flow rate
  • Aggregate removal, Alkaline Stability and Acid cleaning compatibility

Packaging

  • 1.20089.0100: Eshmuno® A Resin 100 ml
  • 1.20089.9010: Eshmuno® A Resin 10 L
  • 1.20089.0010: Eshmuno® A Resin 10 ml
  • 1.20089.0500: Eshmuno® A Resin 500 ml
  • 1.20089.5000: Eshmuno® A Resin 5 L

Plastic bottle

Analysis Note

Appearance: Milky turbid suspension, free from coarse mechanical impurities
Microscopic evaluation: Spherical particles, no agglomerates, no fines
Particle size (30 – 70 µm): ≥ 80 %
Endotoxins: ≤ 1.00 EU/ml
TAMC (Total aerobic microbial count): ≤ 100 CFU/ml
TYMC (Total yeast and mould count): ≤ 100 CFU/ml
Capacity (IgG): ≥ 55 mg/ml
Leakage (Protein A): ≤ 100 ppm

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Multi column chromatography has the benefit of using less resin, increasing productivity, and enabling continuous processing. Learn more in this technical article.

Multi column chromatography has the benefit of using less resin, increasing productivity, and enabling continuous processing. Learn more in this technical article.

Multi column chromatography has the benefit of using less resin, increasing productivity, and enabling continuous processing. Learn more in this technical article.

Multi column chromatography has the benefit of using less resin, increasing productivity, and enabling continuous processing. Learn more in this technical article.

Related Content

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

Learn more on mAb downstream processing, more specifically affinity chromatography and the relevant associated products.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service