Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.20094

Millipore

Eshmuno® P Resin

Eshmuno® P anti-A, 50 μm mean particle size

Synonym(s):

Eshmuno® P anti-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

ligand

(Trisaccharide Blood Group Antigens (A/B))

Quality Level

form

resin

manufacturer/tradename

Eshmuno®

parameter

>500 cm/hr flow rate
8 bar max. pressure

matrix active group

polymer, polyvinyl ether

mean particle size

50 μm

capacity

>75 % binding capacity (anti-A and anti-B)

transition temp

flash point 37.5 °C

application(s)

plasma purification

separation technique

affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Affinity based chromatography resin designed to remove anti-A isoagglutinin antibodies from plasma-derived Ig

Features and Benefits

Eshmuno® P anti-A resin enables users to:

  • Reduce trace amounts of anti- A isoagglutinin′s and the patient risk of hemolysis.
  • Improve the users experience with operational flexibility resulting in high productivity.

Packaging

  • 1.20094.0100: Eshmuno® P anti-A 100 ml
  • 1.20094.0010: Eshmuno® P anti-A 10 ml
  • 1.20094.0500: Eshmuno® P anti-A 500 ml
  • 1.20094.5000: Eshmuno® P anti-A 5 L

Plastic bottle

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

99.5 °F - closed cup - (own results)

Flash Point(C)

37.5 °C - closed cup - (own results)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service