Skip to Content
MilliporeSigma

1.10317

Millipore

Fractogel® EMD BioSEC (S)

Synonym(s):

Fractogel® EMD BioSEC (S)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

ligand

(Modified pore structure)

Quality Level

description

for size exclusion chromatography (SEC), suspension in 20% ethanol and 150 mM NaCl (20-40 µm)

product line

Fractogel®

parameter

100 cm/hr max. flow rate
8 bar pressure

matrix active group

Methacrylate

mean particle size

20-40 μm

transition temp

flash point 35 °C (calculated)

density

1.43 g/cm3 at 20 °C

application(s)

viral therapy

separation technique

size exclusion (SEC)

storage temp.

2-30°C

General description

Size exclusion chromatography for separation of proteins in the range between 5 kDa and 1000 kDa

Features and Benefits

Fractogel® EMD BioSEC (S) enables users to:

  • Effectively remove dimer and high molecular weight aggregates and complete an efficient polishing of blood plasma factors
  • Determine apparent molecular weights of proteins
  • Move predictably and efficiently from bench scale to manufacturing scale processing
  • Create a robust, consistent, and transferable process

Packaging

Plastic bottle

Analysis Note

Appearance: Milky turbid suspension,free from impurities(foreign particles)
Microscopic evaluation: Uniform sphericalparticles, noagglomerates, nofines
Extractable matter (water): ≤ 0.03 %
Cerium: ≤ 1 µg/g
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 5.0 bar
Particle size (d10): 20 - 28 µm
Particle size (d50): 24 - 34 µm
Particle size (d90): 28 - 38 µm
Colony forming units (TAMC + TYMC): ≤ 100 CFU/ml
Endotoxins: ≤ 1.00 EU/ml
Function test (b:a): ≤ 0.7
function test (c:d): ≤ 0.05
Functional Test: Separation of bovine serum albumin, ovalbumin and cytochrom C

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Influenza vaccines are commonly made using egg-based and cell-based manufacturing strategies. Find step-by-step information on the manufacturing process for each method.

A custom-designed cost model is used to explore the economics of vaccine manufacturing across several different modalities including mRNA. The model enables greater process understanding, simulates bottlenecks, and helps to optimize production efficiency.

A custom-designed cost model is used to explore the economics of vaccine manufacturing across several different modalities including mRNA. The model enables greater process understanding, simulates bottlenecks, and helps to optimize production efficiency.

A custom-designed cost model is used to explore the economics of vaccine manufacturing across several different modalities including mRNA. The model enables greater process understanding, simulates bottlenecks, and helps to optimize production efficiency.

See All

Related Content

This technical article breaks down the adenovirus vaccine manufacturing process and provides a case study on developing an accelerated and cost-effective single-use adenoviral vector vaccine.

This technical article breaks down the adenovirus vaccine manufacturing process and provides a case study on developing an accelerated and cost-effective single-use adenoviral vector vaccine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service