Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.12067

Sigma-Aldrich

Lead

fine powder 99+

Synonym(s):

Lead

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
EC Index Number:
231-100-4
NACRES:
NA.22

Quality Level

form

solid

bp

1740 °C/1013 hPa

mp

327 °C

density

11.3 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Analysis Note

Identity (elemental analysis): conforms
Assay (complexometric): ≥ 99.0 %
Particle size (< 100 µm): ≥ 99 %

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service