Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.17952

Supelco

Hydroxymethylfurfural (HMF) Test Strips, reflectometric

1.0-60.0 mg/L (hydroxymethylfurfural (HMF)), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Hydroxymethylfurfural (HMF) chemical test strips, Hydroxymethylfurfural (HMF) test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.05
Pricing and availability is not currently available.

Product Name

Hydroxymethylfurfural (HMF) Test, reflectometric, 1.0-60.0 mg/L (hydroxymethylfurfural (HMF)), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

hydroxymethylfurfural (Hmf)

reaction suitability

reaction type: enzymatic reaction

measuring range

1.0-60.0 mg/L (hydroxymethylfurfural (HMF))

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 1.0 - 60.0 mg/l Reflectoquant®
The freshness of honey is determined by measuring its hydroxymethylfurfural (HMF) content. HMF is an organic compound that arises from the dehydration of fructose, e.g. when honey is heated for easier filling. HMF is barely detectable in freshly centrifuged honey, but increases at 2-3 mg/kg annually, depending on storage temperature and pH. At 21°C, HMF content can rise to 20 mg/kg in just one year. The Reflectoquant HMF Test is the first rapid test for the determination of HMF content, delivering accurate quantitative results in just a few minutes after sample preparation. Easy-to-use, portable and economical, the test is ideal for monitoring raw materials, as well as manufacturing and filling processes. This test provides a simple on-site alternative to time consuming HPLC analysis. On the test strip, HMF reacts with a barbituric acid derivative and an aminophenazone derivative to form a red-violet compound that is determined reflectometrically on the RQflex® 20.

Linkage

Replaces: 1.17952.0007

Analysis Note

function control: conforms

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Hydroxymethylfurfural (HMF) in honey – reflectometric determination after after conversion with a barbituric acid derivative and an aminophenazone derivative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service