Skip to Content
MilliporeSigma

1.12097

Chromium

greener alternative

powder 99+

Synonym(s):

Chromium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
UNSPSC Code:
12141709
EC Index Number:
231-157-5
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

2670 °C/1013 hPa

mp

1890 °C

density

7.20 g/cm3 at 20 °C

greener alternative category

storage temp.

no temp limit

SMILES string

[Cr]

InChI

1S/Cr

InChI key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product falls under category 12 PAP and aligns with the Green Chemistry Principle of Catalysis. Click here for more information.

Analysis Note

Assay (oxidimetric): ≥ 99.0 %
Particle size (< 0.315 mm): ≥ 99


Still not finding the right product?

Explore all of our products under Chromium


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
112097500004027269217136
112097050004022536139319