Skip to Content
MilliporeSigma

1.16128

Malic Acid Test

reflectometric, 5.0-60.0 mg/L (malic acid), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Malic Acid chemical test strips, Malic Acid test strip for instrument reader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Malic Acid Test, reflectometric, 5.0-60.0 mg/L (malic acid), for use with REFLECTOQUANT®

description

quantitative

Quality Segment

specific analyte(s)

malic acid

reaction suitability

reaction type: enzymatic reaction

measuring range

5.0-60.0 mg/L (malic acid)

application(s)

beverage testing
food testing

compatibility

RQflex® 20, for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 5.0 - 60.0 mg/l Reflectoquant®
The Reflectoquant Malic Acid Test can be used for the determination of the decrease in malic acid concentration during the wine-making process. Malic acid (malate) is oxidized to oxalacetate by NAD under the catalytic effect of malate dehydrogenase. In the presence of diaphorase the NADH formed in the process reduces a tetrazolium salt to a red formazan that is determined reflectometrically on the RQflex® 20.

Analysis Note

function control: entspricht
Assay
- standard 1:
6.5 - 8.5 mg/l
- standard 2: 22.5 - 27.5 mg/l
- standard 3: 45.0 - 55.0 mg/l

Other Notes

Replaces: 16128; 16128-1

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
116128000104027269000899
116128000204027269012014