Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.15935

Sigma-Aldrich

Eosin Y (yellowish) (C.I. 45380)

for microscopy Certistain®

Synonym(s):

Eosin Y (yellowish) (C.I. 45380)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
MDL number:
UNSPSC Code:
41116130
EC Index Number:
241-409-6

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

composition

Dye, ≥85% spectrophotometric assay

pH

9.2 (20 °C, 10 g/L in H2O)

mp

295-296 °C

solubility

300 g/L

bulk density

710 kg/m3

εmax

1200-1400 at 515-518 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C20H8Br4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

SEACYXSIPDVVMV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Eosin Y (C.I. 45380) is a Certistain® dry dye for microscopy that is often used as a counterstain, vizualising proteins, connective tissues, fibers and keratin in sample material of human origin. Depending on user requirements and personal preferences, it can be used for the preparation of the commonly used aqueous or alcoholic Eosin Y staining solutions. This staining solution will mostly be used for routine staining in the hematoxylin-eosin staining process (H&E). The H&E staining process is the most frequently used staining method for histological material. Due to the hematoxylin solution, the nuclei will appear blue to dark violet in color and the Eosin Y solution will stain the cytoplasma and intercellular substances red-orange.
As the Eosin Y dye belongs to the portfolio of Certistain® dyes, it is chemically analyzed according to strict specifications before release (CE marked and IVD product) and a high staining quality, with reliable staining results is guaranteed. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (gravimetrically): ≥ 85 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (water): 515 - 518 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): 1200 - 1400
TLC-Test: passes test
Loss on drying (110 °C): ≤ 8 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

376.9 °F - Pensky-Martens closed cup

Flash Point(C)

191.6 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ratchakrit Srikuea et al.
Scientific reports, 10(1), 8239-8239 (2020-05-20)
Skeletal muscle exhibits enormous plasticity throughout life, however, less is known regarding how the stages of growth regulate its local vitamin D system. Herein, we investigated serum 25(OH)D3 and Ca2+ levels along with the vitamin D system in skeletal muscle
Jo Waaler et al.
Communications biology, 3(1), 196-196 (2020-04-26)
The development of immune checkpoint inhibitors represents a major breakthrough in cancer therapy. Nevertheless, a substantial number of patients fail to respond to checkpoint pathway blockade. Evidence for WNT/β-catenin signaling-mediated immune evasion is found in a subset of cancers including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service