Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

199540

Sigma-Aldrich

Ethyl eosin

certified by the BSC, certified by the Biological Stain Commission

Synonym(s):

Eosin alcohol soluble, 2′,4′,5′,7′-Tetrabromoeosin ethyl ester potassium salt, Solvent Red 45

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H11Br4KO5
CAS Number:
Molecular Weight:
714.03
Colour Index Number:
45386
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

Agency

certified by the BSC

form

powder

composition

Dye content, 95%

λmax

532 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[K+].CCOC(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C22H12Br4O5.K/c1-2-30-22(29)10-6-4-3-5-9(10)15-11-7-13(23)18(27)16(25)20(11)31-21-12(15)8-14(24)19(28)17(21)26;/h3-8,27H,2H2,1H3;/q;+1/p-1

InChI key

UKZQEOHHLOYJLY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in the Stoval and Black method for demonstrating Negri bodies in the central nervous system of rabid animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sean Allen et al.
Journal of controlled release : official journal of the Controlled Release Society, 262, 91-103 (2017-07-25)
Flash nanoprecipitation (FNP) has proven to be a powerful tool for the rapid and scalable assembly of solid-core nanoparticles from block copolymers. The process can be performed using a simple confined impingement jets mixer and provides an efficient and reproducible
Analysis and testing of biological stains--the Biological Stain Commission Procedures.
D P Penney et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 77(5-6), 237-275 (2003-02-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service