Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04302

Sigma-Aldrich

Hematoxylin cryst. (C.I. 75290)

for microscopy

Synonym(s):

Hematoxylin cryst. (C.I. 75290)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O6
CAS Number:
Molecular Weight:
302.28
MDL number:
UNSPSC Code:
41116130
EC Index Number:
208-237-3
NACRES:
NA.21

Quality Level

form

solid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

visual transition interval

5.0-7.2, yellow to violet

mp

140 °C (Elimination of water of crystallisation)

bulk density

500 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

O1C[C@@]2([C@H](c4c1c(c(cc4)O)O)c3c(cc(c(c3)O)O)C2)O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m1/s1

InChI key

WZUVPPKBWHMQCE-CJNGLKHVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hematoxylin crystals are a dye for the specific staining of DNA in the nuclei. It is used in cytology and histology. Nuclei are stained blue, dark violet to black. Hematoxylins are mixed with trivalent positively charged metal salts and build up the so-called hematoxylin lakes, used for the selective staining of nuclei (DNA) .
It is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes. For more details, please see instructions for use (IFU).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna-Lena Ruppert et al.
Journal of molecular histology, 51(4), 421-435 (2020-07-04)
Tuft cells are a rare population of chemosensory cells at the mucosal surface epithelia of hollow organs. Their name-giving morphological feature is an apical tuft of stiff microvilli. Accordingly, the actin-binding protein, villin, was identified as one of the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service