Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14750

Supelco

Ammonium Test Kit, colorimetric

0.2-8.0 mg/L (NH₄⁺), MQuant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Ammonium Test, colorimetric, 0.2-8.0 mg/L (NH4+), for use with MCOLORTEST®

specific analyte(s)

ammonium

Quality Level

measuring range

0.2-8.0 mg/L (NH4+)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-disk comparator 0.2 - 0.5 - 0.8 - 1.3 - 2.0 - 3.0 - 4.5 - 6.0 - 8.0 mg/l NH MQuant®
The MQuant® Ammonium test (formally MColortest®) measures both ammonium ions and dissolved ammonia in e.g. groundwater and surface water, drinking water and aquarium water. A pH-dependent equilibrium exists between the two forms.

Method
In strongly alkaline solution ammonia reacts with a chlorinating agent to form monochloramine. This in turn reacts with thymol to form a blue indophenol derivative. Due to the intrinsic yellow coloration of the reagent blank, the measurement solution is yellow-green to green in color. The ammonium concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.

The test is suitable for ammonium determination in
  • Groundwater and surface water
  • Drinking water and mineral water
  • Aquarium water, waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Process water
  • Wastewater
  • Electroplating wastewater
  • Swimming-pool water
  • Nutrient solutions for fertilization
  • Food after appropriate sample pretreatment

This test is not suited for seawater.

Application


  • Evaluation of Water Ammonium Ion Test Kit and Its Feasibility for the Analysis of Protein in Food.: This study assesses a water ammonium ion test kit′s application in food protein analysis, highlighting its potential benefits for ensuring food safety and quality (Wu et al., 2022).

  • Technical note: Evaluation of a colorimetric point-of-care test for measuring urine ammonium concentration in periparturient dairy cattle.: Discusses a colorimetric point-of-care test for measuring urine ammonium in dairy cattle, crucial for animal health monitoring and management (Megahed & Constable, 2020).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service