Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.18387

Supelco

Nitrate Test Kit, colorimetric

5-90 mg/L (NO₃⁻), MQuant®

Synonym(s):

Nitrate Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nitrate Test, colorimetric, 5-90 mg/L (NO3-), for use with MQUANT®

specific analyte(s)

nitrate

Quality Level

measuring range

5-90 mg/L (NO3-)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-disk comparator 5 - 10 - 20 - 30 - 40 - 50 - 60 - 70 - 90 mg/l NO₃⁻ MQuant®
Nitrate is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications. With the MQuant® Nitrate test (formally MColortest®) Nitrates can be closely monitored in:
  • Groundwater, drinking water, and surface water
  • Spring water and well water
  • Mineral water
  • Waters from aquaculture
  • Wastewater and industrial water
  • Nutrient solutions for fertilization
  • Food and beverages after appropriate sample pretreatment
  • Soils after appropriate sample pretreatment
  • Swimming-pool water
This test is not suited for seawater.

Method
In concentrated sulfuric acid nitrate ions react with a benzoic acid derivative to form a red nitro compound. The nitrate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.18462 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Application


  • Soil-test N recommendations augmented with PEST-optimized RZWQM simulations.: This research discusses the use of soil nitrate tests in conjunction with simulation models to optimize nitrogen recommendations for crops, enhancing environmental quality by reducing excess nitrogen use (Malone et al., 2010).

  • Using the late spring nitrate test to reduce nitrate loss within a watershed.: The study focuses on the application of late spring nitrate tests to manage and reduce nitrate leaching in agricultural settings, contributing to improved watershed management (Jaynes et al., 2004).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service