Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.14428

Supelco

Ammonium Test

for use with MQUANT®

Synonym(s):

Ammonium Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

specific analyte(s)

ammonium

Quality Level

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Méthode colorimétrique, bleu d′indophénol, avec carte des couleurs et bloc comparateur avec des tubes à 0.025-0.050-0.075-0.10-0.15-0.20-0.25-0.30-0.40 mg/l NH₄⁺ 0.02-0.04-0.06-0.08-0.12-0.16-0.19-0.23-0.31 mg/l NH₄-N MQuant®

Application


  • Evaluation of Water Ammonium Ion Test Kit and Its Feasibility for the Analysis of Protein in Food.: This study assesses a water ammonium ion test kit′s application in food protein analysis, highlighting its potential benefits for ensuring food safety and quality (Wu et al., 2022).

  • Technical note: Evaluation of a colorimetric point-of-care test for measuring urine ammonium concentration in periparturient dairy cattle.: Discusses a colorimetric point-of-care test for measuring urine ammonium in dairy cattle, crucial for animal health monitoring and management (Megahed & Constable, 2020).

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service