Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03428

Sigma-Aldrich

Deuterium oxide

deuteration degree min. 99.95% for NMR spectroscopy MagniSolv

Synonym(s):

Deuterium oxide, Heavy water

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
MDL number:
UNSPSC Code:
12142201
EC Index Number:
232-148-9
NACRES:
NA.22
grade:
NMR grade
technique(s):
NMR: suitable
bp:
101 °C/1013 hPa

grade

NMR grade

Quality Level

isotopic purity

≥99.95 atom % D

form

liquid

technique(s)

NMR: suitable

pH

7 (20 °C in H2O)

bp

101 °C/1013 hPa

mp

3.8 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O([2H])[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Application


  • Sensing the Bactericidal and Bacteriostatic Antimicrobial Mode of Action Using Raman Deuterium Stable Isotope Probing (DSIP) in Escherichia coli.: This research demonstrates a novel application of Raman spectroscopy enhanced by Deuterium oxide to discern antimicrobial mechanisms (Karlo et al., 2024).


  • Dependence on relative humidity in the formation of reactive oxygen species in water droplets.: Investigates how relative humidity influences the generation of reactive oxygen species in water droplets, using analytical techniques that include Deuterium oxide (Mofidfar et al., 2024).


  • Hydrophilic Interaction Liquid Chromatography-Hydrogen/Deuterium Exchange-Mass Spectrometry (HILIC-HDX-MS) for Untargeted Metabolomics.: This study presents a method integrating Deuterium oxide for untargeted metabolomic analyses, providing a comprehensive approach to biochemical analysis (Cajka et al., 2024).


  • Blood compatibility of poly(propylene glycol diester) and its water structure observed by differential scanning calorimetry and (2)H-nuclear magnetic resonance spectroscopy.: Explores the interaction between polymers and water, where Deuterium oxide plays a crucial role in enhancing the sensitivity of nuclear magnetic resonance spectroscopy (Mochizuki et al., 2024).


Analysis Note

Deuteration degree: ≥ 99.95 %
Performance test (NMR-spectrum): passes test

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service