Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.10083

Supelco

Calcium Test Strips, colorimetric

10-100 mg/L (Ca²⁺), MQuant®

Synonym(s):

Calcium chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Calcium Test, colorimetric, 10-100 mg/L (Ca2+), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

calcium

measuring range

10-100 mg/L (Ca2+)

detection method

colorimetric

storage temp.

2-8°C

General description

Method: Colorimetric with test strips and reagents 10 - 25 - 50 - 100 mg/L (Ca2+) MQuant®

The MQuant® Calcium test includes test strips and all necessary additional reagents and is suitable for rapid determination of calcium in aqueous media. In the presence of hydrogen peroxide calcium ions react with glyoxal-bis(2-hydroxyanil) to form a red complex. The calcium concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. For a quantitative determination of calcium in different concentration ranges please refer to our Reflectoquant® and Spectroquant® tests.

Application


  • Development of a blood calcium test for hypocalcemia diagnosis in dairy cows.: This study focuses on creating a blood calcium test to diagnose hypocalcemia in dairy cows. The test′s development and validation are crucial for improving animal health and productivity in the dairy industry (Fu et al., 2022).


Linkage

Replaces: 10083-1

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service