Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.16993

Supelco

Calcium Test for RQflex® 10 and RQflex® 10 plus

2.5-45.0 mg/L (Ca), REFLECTOQUANT®

Synonym(s):

Calcium for RQflex® 10 and RQflex® 10 plus chemical test strips, Calcium for RQflex® 10 and RQflex® 10 plus test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Calcium Test for RQflex® 10 and RQflex® 10 plus, reflectometric, 2.5-45.0 mg/L (Ca), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

calcium

measuring range

2.5-45.0 mg/L (Ca)

compatibility

RQflex® 10 Plus
RQflex® 10
RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

15-25°C

General description

Method: reflectometric with test strips and reagents 2.5 - 45.0 mg/l Ca Reflectoquant®
The Reflectoquant Calcium test includes test strips and all necessary additional reagents and is suitable for rapid determination of calcium in aqueous media. Calcium is an important parameter for boiling and cooling water. If the water contains calcium, deposits can be found in the plants and this can influence the function of the machines. Calcium also needs to be analyzed in drinking water and especially in beer. In the presence of hydrogen peroxide calcium ions react with glyoxal-bis(2-hydroxyanil) to form a red complex that is determined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the calcium concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for calcium. Further quantitative determination methods can be found with our Spectroquant tests.

Analysis Note

function control: conforms
Assay
- standard 1:
4.4 - 5.6 mg/l
- standard 2: 13.5 - 16.5 mg/l
- standard 3: 25.0 - 31.0 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service