Skip to Content
MilliporeSigma

1.10004

Iron Test Strips, colorimetric

greener alternative

3-500 mg/L (Fe²⁺), MQuant®

Synonym(s):

Iron chemical test strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Iron Test, colorimetric, 3-500 mg/L (Fe2+), MQuant®

description

semi-quantitative

Quality Segment

product line

MQuant®

specific analyte(s)

iron

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

3-500 mg/L (Fe2+)

application(s)

beverage testing
drinking water
food testing
ground and surface water
milk and dairy products
wastewater

compatibility

Reflectoquant®, Spectroquant®

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips, 3 - 10 - 25 - 50 - 100 - 250 - 500 mg/L (Fe²+), MQuant®

The MQuant® test strips offer a very simple means of determining iron(II) in all aqueous media and on metal surfaces. They can thus be used to distinguish between ferrous and non-ferrous metals. Iron(II) ions react with 2,2′-bipyridine to form a red complex. The iron(II) concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. For a determination of Fe(III) it is necessary to treat the sample with Ascorbic acid first, to reduce Fe(III) to Fe(II). The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the iron concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® test.

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.This product has benefited from:
  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Other Notes

Replaces: 10004-1; 10004

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library