Skip to Content
MilliporeSigma

1.17922

Peracetic Acid Test Strips, colorimetric

500-2000 mg/L (peracetic acid), MQuant®

Sustainability features

Synonym(s):

Peracetic Acid chemical test strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
eCl@ss:
40050204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Peracetic Acid Test, colorimetric, 500-2000 mg/L (peracetic acid), MQuant®

description

semi-quantitative

Quality Segment

product line

MQuant®

specific analyte(s)

peracetic acid

reaction suitability

reaction type: Redox Reactions

sustainability

Sustainability features

measuring range

500-2000 mg/L (peracetic acid)

application(s)

cannabis testing
disinfection control

detection method

colorimetric

storage temp.

2-8°C

General description

Method: Colorimetric with test strips, 500 - 1000 - 1500 - 2000 mg/L, MQuant®

Peracetic acid is a frequently used disinfectant. Due to its measuring range the MQuant® Peracetic acid test strips are especially suitable to check the presence of effective levels in peracetic acid based disinfectants. Peracetic acid reacts with a phenol derivative to form a violet dye. The concentration of peracetic acid is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the peracetic acid concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip width reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Peracetic Acid Test


Storage Class

11 - Combustible Solids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library