Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09491

Supelco

ICP multi-element standard solution XI

for sludge testing (7 elements in dilute nitric acid) Certipur®

Synonym(s):

Nota 1 A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

density

1.045 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Analysis Note

Cd (cadmium): 10 mg/l
Cr (chromium): 900 mg/l
Cu (Copper): 800 mg/l
Hg (Mercury): 8 mg/l
Ni (Nickel): 200 mg/l
Pb (lead): 900 mg/l
Zn (Zinc): 2500 mg/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service