Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43843

Supelco

Multielement Standard Solution 6 for ICP

TraceCERT®, 23 elements, 100 mg/L each in nitric acid and hydrofluoric acid

Synonym(s):

ICP Multi-Element Calibration Solution 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

technique(s)

AAS: suitable
ICP: suitable

application(s)

industrial qc
pharmaceutical

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Multielement standard solution 6 can be used as a calibration standard in the:
  • Determination of trace metals in petroleum cokes using inductively coupled plasma optical emission spectrometry system (ICP-OES).
  • Trace metal analysis of seawater and fish otoliths via ICP-MS, enhancing the detection and quantification of trace metals critical in environmental samples.

Analysis Note

100 mg/L: Al, Sb, Ba, Pb, B, Ca, Cd, Cr, Co, Fe, K, Cu, Li, Mg, Mn, Mo, Na, Ni, P, Si, Ti, V, and Zn in 5% nitric acid and ≤0.1 % hydrofluoric acid.

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service